Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N012
Encompass_Preferred
The Grainger 16N012 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing, Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed, fan cooled (TEFC) with a cast iron frame and end bells. It has Class F insulation and is rated for continuous duty applications.
Catalog No. 50-276-6066