Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N022
Encompass_Preferred
The Grainger 16N022 is a 3/4 HP, 115V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, Ball Bearing Electric Motor. It has a NEMA standard 56C frame with a 1.15 service factor. The motor is totally enclosed and fan cooled with Class F insulation and a C-face endshield. It can provide 0.56 kW of output power. The 16N022 would be a good choice for powering pumps, fans, conveyors, compressors, machine tools, or other industrial equipment needing up to 3/4 horsepower.
Catalog No. 50-276-6067