Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N060
Encompass_Preferred
The Grainger 16N060 is a 16 oz. dead blow plastic hammer with a 2-1/2 inch diameter striking face. It has a hollow head filled with steel and lead shot to prevent rebound and absorb shock. The hammer has a fiberglass handle that provides durability. This hammer allows the user to deliver heavy blows without marring the surface and is well-suited for assembling delicate materials or working in tight spaces. The hammer weighs approximately 1.1 lbs.
Catalog No. 50-276-8363