Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N046
Encompass_Preferred
The Gasco Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and testing. It comes in a 34L cylinder with CGA 580 inlet connection. The gas purity is 99.998% nitrogen. The cylinder operating pressure is 2000 psig with a capacity of 103L. This calibration gas meets standards for use in calibration and testing of gas detection and analysis equipment. The product number for ordering is 16N026. This is a useful calibration gas for precise testing and calibration using pure nitrogen.
Catalog No. 50-276-5922