Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N027
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing air for calibrating gas detection instruments. This 16 liter aluminum cylinder contains 99.9995% pure air certified to NIST traceable standards. The cylinder has a CGA-590 outlet with a brass valve and is refillable. This calibration gas is suitable for calibrating and bump testing gas detectors used for monitoring oxygen levels. With a 2 year shelf life, this cylinder from GASCO provides high purity air from a reliable supplier. The 16N027 model contains 16 liters of gas at a pressure of 1800 psi.
Catalog No. 50-276-4987