Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N029
Encompass_Preferred
The GASCO Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and testing of gas detection equipment. It comes in a 103L cylinder with 99.998% nitrogen. The cylinder has a CGA 580 outlet connection and is rated for service pressure up to 3000 psig. This calibration gas meets ISO 6141 standards and is traceable to NIST standards. The nitrogen gas has no odor and is nonflammable, nontoxic and inert. It can be used for purging lines, leak testing equipment and calibrating gas detectors. The cylinder must be secured upright during use and storage. Proper PPE should be worn when handling high pressure gas cylinders.
Catalog No. 50-276-5048