Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N020
Encompass_Preferred
The Grainger 16N020 is a 16 oz. aerosol can of CRC Brakleen Brake Parts Cleaner. It is a fast drying, non-flammable brake parts cleaner that removes oil, grease, brake fluid, and other contaminants from brake linings and drums. It leaves no residue and can be used on metallic and non-metallic brake parts. The cleaner sprays out as a liquid and features a 360° valve that allows spraying in any position.
Catalog No. 50-276-7234