Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N038
Encompass_Preferred
The GASCO Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and testing of gas detection equipment. It comes in a 34L aluminum cylinder with CGA 580 outlet connection. The gas purity is 99.998% nitrogen. The cylinder operating pressure is 1800 psig. This calibration gas meets NIST traceability standards and is ISO 6141 compliant. The 17 digit part number for this product is 16N028.
Catalog No. 50-276-7644