Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N037
Encompass_Preferred
The Gasco Calibration Gas Nitrogen is a high purity nitrogen gas used for calibration and testing of gas detection equipment. It comes in a 103L cylinder with 99.998% nitrogen. The cylinder has a CGA 580 outlet connection and is rated for pressures up to 3000 psig. This calibration gas meets ISO 6141 standards and is traceable to NIST standards. The gas has a two year recommended shelf life. This nitrogen gas cylinder is suitable for calibrating and bump testing gas detectors, calibration benches, and other instrumentation.
Catalog No. 50-276-5897