Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N006
Encompass_Preferred
The Gasco Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and testing of gas detection equipment. It comes in a 103L cylinder with 99.998% nitrogen. The cylinder has a CGA 580 outlet connection and is rated for service pressure up to 3000 psig. This calibration gas allows for accurate testing and calibration of gas detectors, analyzers, and other instrumentation. The high purity nitrogen can verify proper operation and response factors. With proper handling and storage, the shelf life is 2 years from the date of fill. This nitrogen gas cylinder is an ideal calibration tool for industrial safety applications.
Catalog No. 50-276-7028