Learn More
Grainger Calibration Gas 116L 3-Gas Mix
Supplier:  Grainger 16N014
Encompass_Preferred
The Gasco Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and purging. It has a purity of 99.998% and comes in a 103L cylinder with CGA 580 inlet and outlet connections. The cylinder has a working pressure of 2000 psig. Nitrogen is nonflammable, noncorrosive, and inert. It can be used for purging lines and equipment of flammable gases, inerting storage tanks, and calibrating gas analyzers and detectors. This cylinder of nitrogen gas is manufactured under strict quality control standards for guaranteed purity and performance.
Catalog No. 50-276-6134