Learn More
Grainger Calibration Gas 116L Ammonia Air
Supplier:  Grainger 16N134
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing air for calibrating gas detection instruments. This 34 liter aluminum cylinder contains 99.9995% pure air balanced with nitrogen. It has a working pressure of 1800 psi and comes with a CGA-590 outlet connection that is compatible with many gas detection instruments. The cylinder should be stored and used in accordance with local regulations. This calibration gas is useful for verifying calibration on gas detectors used for oxygen deficiency monitoring.
Catalog No. 50-276-6047