Learn More
Grainger Calibration Gas 116L Carbon Dioxide Air
Supplier:  Grainger 16N071
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing compressed air. It has a capacity of 103L and is made of aluminum. The cylinder has a CGA-590 outlet connection that is compatible with regulators and flowmeters. The air meets ASTM D1142, ISO 6141, and SRM 2890 specifications and is certified to contain 20.9% oxygen. It can be used for calibration and bump testing of gas detection instruments. The cylinder is refillable and requires a CGA-590 fill adapter. It is suitable for calibration of a wide variety of gas detection and measurement instruments.
Catalog No. 50-276-6024