Learn More
Grainger Calibration Gas 116L Carbon Monoxide Air
Supplier:  Grainger 16N081
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas used to calibrate gas detection instruments. It contains 20.9% oxygen with the balance being nitrogen. The gas comes in a 34 liter cylinder with a CGA-580 outlet connection. The cylinder operating pressure is 1800 psi. This calibration gas meets NIST traceability standards and has a shelf life of 36 months. It can be used to calibrate and bump test gas detectors used for monitoring oxygen levels. This is a high purity gas mixture suitable for calibrating and testing air quality monitoring equipment.
Catalog No. 50-276-8429