Learn More
Grainger Calibration Gas 116L Chlorine Nitrogen
Supplier:  Grainger 16N142
Encompass_Preferred
The IDS 18101758 Cylinder Calibration Gas 10ppm Chlorine is a high-purity gas mixture used for calibration and bump testing of fixed and portable chlorine gas detectors. It contains 10 parts per million of chlorine gas balanced with nitrogen. The gas comes in a 34L aluminum cylinder with CGA 590 outlet connection. The cylinder has a working pressure of 1800 psi. This calibration gas is ideal for verifying chlorine sensor response and checking that your gas detectors are working properly. It has a shelf life of 18 months and is manufactured in the USA.
Catalog No. 50-276-4925