Learn More
Grainger Calibration Gas 116L Ethylene Air
Supplier:  Grainger 16N088
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing air for calibrating gas detection instruments. This 34 liter aluminum cylinder contains 99.9995% pure air balanced with nitrogen and is certified to NIST traceable standards. The cylinder has a CGA-590 outlet with a brass valve and is refillable. This calibration gas is suitable for calibrating and bump testing gas detectors used for monitoring oxygen levels. With a 2 year shelf life, this cylinder from GASCO provides high purity air from a reliable gas supplier. The 16N088 model contains air at approximately 2000 psi fill pressure.
Catalog No. 50-276-6076