Learn More
Grainger Calibration Gas 116L Ethylene Air
Supplier:  Grainger 16N089
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing air for calibrating gas detection instruments. This 34 liter aluminum cylinder contains 99.9995% pure air balanced with nitrogen. It has a maximum allowable working pressure of 1800 psi at 70°F and comes with a CGA-590 outlet connection that is compatible with many gas detection instruments. The cylinder should be stored and used in accordance with local regulations. This calibration gas is suitable for calibrating and bump testing many different gas detectors to ensure they are functioning properly.
Catalog No. 50-276-8511