Learn More
Grainger Calibration Gas 116L Hexane Air
Supplier:  Grainger 16N102
Encompass_Preferred
The Gasco Calibration Gas Nitrogen is a high purity nitrogen gas used for calibration and testing of gas detection equipment. It comes in a 34L aluminum cylinder with CGA 580 outlet connection. The gas purity is 99.998% nitrogen. The cylinder operating pressure is 1800 psig at 70°F. This calibration gas meets NIST traceability standards and is ISO/IEC 17025 accredited. The 34L cylinder contains approximately 3400 liters of gas at atmospheric pressure. This is an ideal nitrogen gas source for span gas calibration of gas analyzers and detectors.
Catalog No. 50-276-5836