Learn More
Grainger Calibration Gas 116L Methane Air
Supplier:  Grainger 16N114
Encompass_Preferred
The GASCO 116ES-150A-10 Calibration Gas 116L is a 150 liter cylinder of 10 parts per million methane in air calibration gas mixture. It is certified to NIST traceable standards and is ideal for calibrating methane gas detectors. The high pressure cylinder has a CGA-590 outlet valve and is rated for pressures up to 3000 psig. The gas mixture has a shelf life of 18 months and is available for fast delivery directly from the manufacturer GASCO.
Catalog No. 50-276-7017