Learn More
Grainger Calibration Gas 116L Methane Nitrogen
Supplier:  Grainger 16N115
Encompass_Preferred
The Gasco Calibration Gas Methane is a high purity methane gas mixture used for calibration and bump testing of gas detection instruments. It contains 1000 ppm methane, with nitrogen as the balance gas, and is available in a 104 liter cylinder with CGA-580 outlet connection. This calibration gas has an accuracy of ±2% and is recommended for calibration of combustible gas detectors. The cylinder operating pressure is 1800 psi at 70°F.
Catalog No. 50-276-7018