Learn More
Grainger Calibration Gas 116L Methane Air
Supplier:  Grainger 16N109
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing air for calibrating gas detection instruments. It has a capacity of 103L and contains 20.9% oxygen with the balance being nitrogen. The cylinder has a CGA-590 outlet connection and comes in an aluminum cylinder that is 49” tall x 8” diameter. The product has a 2 year shelf life and is manufactured in the USA. It can be used for calibrating and bump testing gas detectors to ensure proper operation and safety. The cylinder is disposable and not refillable.
Catalog No. 50-276-5627