Learn More
Grainger Calibration Gas 116L Oxygen Nitrogen
Supplier:  Grainger 16N122
Encompass_Preferred
The Grainger 16N122 is a 16 oz. spray bottle made of HDPE plastic with a 28/400 neck finish. It has an ergonomic trigger handle and a 6 inch hose for easy dispensing. This bottle can hold liquids up to 160°F and has a max pressure of 60 PSI. It features an adjustable cone and stream spray nozzle. The bottle measures 10-1/2" tall x 3" wide and has volume markings up to 16 oz. This general purpose spray bottle is suitable for use with cleaning chemicals, lubricants, pesticides and other industrial liquids. It meets FDA and USDA standards for chemical compatibility. The bottle is translucent to show fluid level.
Catalog No. 50-276-7363