Learn More
Grainger Calibration Gas 116L Pentane Air
Supplier:  Grainger 16N126
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing compressed air. It has a capacity of 103L and is made of aluminum. The cylinder has a CGA-590 outlet connection that is compatible with regulators and flowmeters. The air meets ASTM D1142, ISO 6141, and SRM 2890 specifications and is certified to contain 20.9% oxygen. It can be used for calibration and bump testing of gas detectors and analyzers. The cylinder is refillable and is marked with a 10 year requalification date from the manufacture date. It requires a CGA-590 compatible regulator to dispense the gas.
Catalog No. 50-276-6484