Learn More
Grainger Calibration Gas 116L Pentane Air
Supplier:  Grainger 16N125
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing zero air. It has a capacity of 103L and is made of aluminum. This cylinder of zero air can be used for calibration and bump testing of gas detectors and monitoring systems. The cylinder has a CGA 590 outlet connection and comes equipped with a brass 3-way valve. The gas mixture is 21% oxygen and 79% nitrogen. This calibration gas cylinder allows for accurate sensor testing and calibration. The cylinder is refillable and meets ISO standards. It is suitable for calibrating and bump testing a variety of gas detection equipment.
Catalog No. 50-276-5668