Learn More
Grainger Calibration Gas 116L Phosphine Nitrogen
Supplier:  Grainger 16N171
Encompass_Preferred
The Gasco Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and purging. It has a purity of 99.998% and comes in a 103L cylinder with CGA 580 outlet connection. The cylinder operating pressure is 2000 psig. This calibration gas meets standards for instrumental gas analysis and is ideal for use in laboratories. The product number is 16N171. This is a high quality nitrogen gas for all calibration and purging needs.
Catalog No. 50-276-5838