Learn More
Grainger Calibration Gas 116L Propane Air
Supplier:  Grainger 16N128
Encompass_Preferred
The GASCO Calibration Gas - Air is a calibration gas cylinder containing compressed air. It has a capacity of 103L and is made of aluminum. The cylinder has a CGA-590 outlet connection that is compatible with regulators and flowmeters. The gas mixture is 20.9% oxygen and 79.1% nitrogen. It can be used for calibration and bump testing of gas detection instruments. The cylinder is DOT approved and comes with a certificate of analysis from the manufacturer specifying the gas concentrations. The product has a 12 month shelf life and is available in high pressure cylinder sizes of 34L, 58L, 103L. It requires the use of proper regulators, gauges, and flow control equipment.
Catalog No. 50-276-7329