Learn More
Grainger Calibration Gas 116L R11 Nitrogen
Supplier:  Grainger 16N094
Encompass_Preferred
The Gasco Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and purging. It has a purity of 99.998% and comes in a 103L cylinder with CGA 580 outlet connection. The gas mixture is non-flammable and non-toxic. It can be used for purging lines and equipment of reactive gases or calibrating gas analyzers and detectors. This nitrogen calibration gas is ideal for laboratories and industrial facilities that require pure nitrogen gas for instrumentation.
Catalog No. 50-276-8512