Learn More
Grainger Calibration Gas 116L R12 Nitrogen
Supplier:  Grainger 16N096
Encompass_Preferred
The GASCO Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and testing of gas detection equipment. It comes in a 103L cylinder with 99.998% nitrogen. The cylinder has a CGA 580 outlet connection and is rated for service pressure up to 3000 psig. This calibration gas meets standards for instrument calibration gas and provides accurate, repeatable results when used for routine maintenance. The cylinder is refillable and is a reliable source of nitrogen gas for calibration and testing needs.
Catalog No. 50-276-8552