Learn More
Grainger Calibration Gas 116L R22 Nitrogen
Supplier:  Grainger 16N093
Encompass_Preferred
The GASCO Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and testing of gas detection equipment. It comes in a 103L cylinder with 99.998% nitrogen. The cylinder has a CGA 580 outlet connection and pressure of 2000 psig. The product has a 2 year recommended shelf life. This nitrogen gas meets standards for calibration and bump testing of gas detectors and analyzers. It can be used for purging lines and as a shield gas. The cylinder is refillable.
Catalog No. 50-276-7564