Learn More
Grainger Calibration Gas 116L Silane Nitrogen
Supplier:  Grainger 16N172
Encompass_Preferred
The GASCO Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and purging. It has a purity of 99.998% and comes in a 116 liter cylinder with a CGA 580 outlet connection. The cylinder operating pressure is 2000 psig. This calibration gas meets standards for use in calibration and testing of gas detection and analysis equipment. The product number for this item is 16N116. This nitrogen gas is useful for providing an inert atmosphere and baseline reference for calibration.
Catalog No. 50-276-5333