Learn More
Grainger Calibration Gas 116L Toluene Nitrogen
Supplier:  Grainger 16N132
Encompass_Preferred
The Gasco Calibration Gas - Nitrogen is a high purity nitrogen gas used for calibration and purging. It has a purity of 99.998% and comes in a 116 liter cylinder with a CGA 580 outlet connection. The cylinder operating pressure is 2000 psig. This calibration gas meets National Institute of Standards and Technology standards and is ideal for calibrating gas analyzers and monitoring systems. The product number is 16N116.
Catalog No. 50-276-6532