Learn More
Spherotech CALIBRATION PARTICLES
Supplier:  Spherotech ACURFP2.53001
Encompass_Preferred
AccuCount UltraRainbow Calibration Particles, 2.5x106/mL, 30.0 um (+/-2 um), 1 mL
Catalog No. NC2108240