Learn More
Unchained Labs High Lunatic Plates, OD 200
SDP
Supplier:  Unchained Labs 7012000
Encompass
Box of 10 High Lunatic Plates, 0.03-200 OD. 96 samples per plate. For use with Big Lunatic, DropSense96 or DropSense96 Classic. (Previously product code 4-210096-D+)
Catalog No. 50-112-5955