Learn More
Cayman Chemical CalIcheamIcIn 1 10mg
SDP
Supplier:  Cayman Chemical 4083010 MG
Encompass_Preferred
A polyketide enediyne; induces double-stranded DNA scission in cell-free assays
Catalog No. 50-294-3341