Learn More
Abcam Calmidazolium chloride, Calmodulin inhibitor, 10MG
Supplier:  Abcam AB12065810MG
Encompass_Preferred
MW 687.7 Da, Purity >98%. Calmodulin inhibitor (IC₅₀ = 10 nM). Elevates intracellular calcium, independent of calmodulin inhibition. Additionally inhibits adenylyl cylase activity.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7379