Learn More
Medchemexpress LLC Caloxin 1C2 | 99.5% | 1844.08 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYP10655
Encompass_Preferred
Caloxin 1C2 is a specific plasma-membrane Ca2+-ATPases (PMCA) inhibitor. It exhibits selectivity for PMCA4 over PMCA1, 2, and 3, and does not significantly affect total Ca2+ transfer. This inhibitor also impacts coronary contractility.
  • Specific PMCA inhibitor
  • Selective for PMCA4 over PMCA1, 2, and 3
  • Does not significantly affect total Ca2+ transfer
  • Affects coronary contractility
  • Ki for PMCA4 is 2.3 ± 0.3 μM
  • Ki for PMCA1 is 21 ± 6 μM
Catalog No. 50-004-25467