Learn More
Cayman Chemical CalpaIn InhIbItr III 25mg
SDP
Supplier:  Cayman Chemical 1428325 MG
Encompass_Preferred
A cell permeable, selective inhibitor of μ-calpain (calpain-1) and m-calpain (calpain-2); crosses the blood-brain barrier to inhibit brain cysteine protease activity; reported to have protective effects in numerous rodent models of neurotrauma and cardiac ischemia
Catalog No. 50-295-4136