Learn More
Cusabio Technology LLC Pig calprotectin (CALP) ELISA kit
Supplier:  Cusabio Technology LLC CSBEQ013485PI
Encompass_Preferred
This Pig CALP ELISA Kit was designed for the quantitative measurement of Pig CALP protein in serum, plasma, tissue homogenates. It is a Competitive ELISA kit, its detection range is 0.312 ng/mL-20 ng/mL and the sensitivity is 0.078 ng/mL. One kit contains 96 wells.
Catalog No. NC2952989