Learn More
Alpco Ltd Calprotectin Chemiluminescence ELISA, 15 mg, 96 Wells
SDP
Supplier:  Alpco Ltd 80CALPHUCH01
Encompass_Preferred
The ALPCO Calprotectin Chemiluminescence ELISA is an in vitro diagnostic chemiluminescent assay intended for the quantitative measurement of fecal calprotectin, a neutrophilic protein that is a marker of intestinal mucosal inflammation, in human stool. The ALPCO Calprotectin Chemiluminescence ELISA is intended for in vitro diagnostic use as an aid in the diagnosis of inflammatory bowel disease (IBD), specifically Crohn’s disease (CD) and ulcerative colitis (UC), and as an aid in the differentiation of IBD from irritable bowel syndrome (IBS) in conjunction with other clinical and laboratory findings.
The Calprotectin Chemiluminescence ELISA is the most clinically accurate test on the market and provides the following:
  • The best combination of clinical specificity (95.1%) and clinical sensitivity (92.1%) when used as an aid in the differentiation of IBD from IBS.
Catalog No. NC2233090