Learn More
Medchemexpress LLC Calreticulin/CALR M 10ug

Supplier: Medchemexpress LLC HYP7211210UG
Calreticulin (CALR) Protein, Mouse (GST) is a recombinant, full-length mature mouse calreticulin expressed in E. coli and supplied lyophilized. The protein includes an N-terminal GST affinity tag, is >95% pure, and is formulated for stability, making it suitable for biochemical assays, structural studies, and antibody validation.
- Full-length mature sequence (D18-L416).
- N-terminal GST tag for affinity purification.
- Expressed in E. coli to enable high-yield production.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Lyophilized formulation with trehalose for enhanced stability.
- Store at -20°C; reconstitute and aliquot following supplier guidance.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.