Learn More
Medchemexpress LLC Calreticulin/CALR M 10ug

Supplier: Medchemexpress LLC HYP71721Y10UG
Recombinant mouse calreticulin (CALR) is a calcium-binding molecular chaperone supplied as a filtered solution in PBS with 50% glycerol. Expressed in Pichia pastoris with an N-terminal 6xHis tag, this preparation is intended for biochemical assays, protein-protein interaction studies, and ER/chaperone research requiring high-purity, stable protein.
- Expressed in Pichia pastoris for eukaryotic post-translational processing.
- Contains an N-terminal 6xHis tag to facilitate purification and detection.
- Supplied as a 0.22 μm filtered PBS solution (pH 7.4) containing 50% glycerol for stability.
- Purity greater than 90% as determined by reducing SDS-PAGE.
- Approximate molecular weight 48.3 kDa.
- Stable at -80°C for long-term storage; avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.