Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Calreticulin/CALR M 50ug
SDP

Supplier:  Medchemexpress LLC HYP7211250UG

Encompass_Preferred

Recombinant mouse calreticulin (GST-tagged) is a calcium-binding molecular chaperone supplied lyophilized for biochemical, binding, and assay development applications. The protein is expressed in E. coli and formulated for stability and low endotoxin to support sensitive downstream experiments.

  • Expressed in E. coli for high-yield production.
  • N-terminal GST tag for purification and detection.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
  • Lyophilized from Tris-HCl with trehalose for enhanced stability.
  • Store at -20°C; aliquot after reconstitution to avoid repeated freeze-thaw.

Catalog No. 50-001-95379


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.