Learn More
Medchemexpress LLC Calreticulin/CALR M 5ug

Supplier: Medchemexpress LLC HYP717215UG
Recombinant full-length mouse calreticulin (CALR) expressed in Pichia pastoris and supplied as an N-terminal 6xHis-tagged soluble protein in PBS with 50% glycerol. The product is provided at high purity with low endotoxin and is intended for biochemical and molecular biology research such as binding, structural, and functional assays.
- Recombinant full-length mouse protein for biochemical and molecular biology use.
- Expressed in Pichia pastoris to support eukaryotic folding and modifications.
- N-terminal 6xHis tag facilitates detection and affinity purification.
- Supplied as a 0.22 μm filtered PBS solution with 50% glycerol for stability.
- Purity greater than 90% by reducing SDS-PAGE.
- Low endotoxin level (<1 EU/μg) suitable for sensitive assays.
- Available in small quantities, including 5 μg, for low-volume experiments.
- Shipped on dry ice; store at -80°C and avoid repeated freeze-thaw cycles.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.