Learn More
Cell Signaling Technology Calreticulin (D3E6) XP Rabbit mAb (Alexa Fluor 555 Conjugate), 100 uL
SDP
Supplier:  Cell Signaling Technology 52286S
Encompass
This Cell Signaling Technology antibody is conjugated to Alexa Fluor 555 fluorescent dye and tested in-house for direct flow cytometric analysis in human cells and immunofluorescent analysis in mouse and human cells. This antibody is expected to exhibit the same species cross-reactivity as the unconjugated Calreticulin (D3E6) XP Rabbit mAb #12238.
Catalog No. NC1698954