Learn More
Medchemexpress LLC Calycosin-7-O-β-D-glucoside | 20633-67-4 | MFCD08061493 | 446.40 | 1 ML

Supplier: Medchemexpress LLC HYN052010MM/1ML
Calycosin-7-O-β-D-glucoside is an isoflavone isolated from Astragali Radix. It possesses various biological activities, including neuroprotective, cardioprotection, anti-inflammatory, and antioxidative stress effects.
- Neuroprotective effects
- Cardioprotection
- Anti-inflammation
- Antioxidative stress effects
- MMPs inhibition
- BBB integrity
- Anticancer activity
- Purity: 99.05%
- Appearance: Solid, white to off-white
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.