Learn More
Enzo Life Sciences CaM kinase 2 (290-309) (0.5mg). CAS: 115044-69-4
SDP
Supplier:  Enzo Life Sciences BMLP2000500
Encompass_Preferred
Synthetic peptide with sequence from Ca2+/calmodulin-dependent protein kinase II pseudosubstrate site. Peptide binds calmodulin tightly and inhibits calmodulin-requiring enzymes including CaMKII, phosphodiesterase and calcineurin. IC50=52 nM for CaMKII in the presence of 100 nM calmodulin. Alternative name: CaMK2 (290-309), Ca2+ / calmodulin dependent protein kinase II (290 309). Purity: ≥97%. Long Term Storage: -20°C.
Catalog No. 50-201-1277