Learn More
Medchemexpress LLC HY-119502 5mg Medchemexpress, Camalexin CAS:135531-86-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1195025MG
Encompass_Preferred
Medchemexpress, HY-119502 5mg Camalexin CAS:135531-86-1 Camalexin is a phytoalexin isolated from Camelina sativa and Arabidopsis (Cruciferae) with antibacterial, antifungal, antiproliferative and anticancer activities. Camalexin can induce reactive oxygen species (ROS) production[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9540