Learn More
Cayman Chemical Cambendazol 10mg
SDP
Supplier:  Cayman Chemical 2084110 MG
Encompass_Preferred
An antihelmintic agent; eradicates S. ratti adult worms from intestine and S. stercoralis larva from muscle at a dose of 50 mg/kg/day; inhibits polymerization of microtubules isolated from bovine brain (IC50 = 64.2 µM)
Catalog No. 50-298-2414