Learn More
Abcam Cambinol, SIRT1/2 inhibitor, 25MG
Supplier:  Abcam AB14118125MG
Encompass_Preferred
MW 360.4 Da, Purity >98%. SIRT1/2 inhibitor (IC₅₀ values are 56 and 59 μM at SIRT1 and SIRT2 respectively). Class III HDAC inhibitor, with no inhibition of class I and II. Promotes G₂ cell cycle arrest. Antitumour activity in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6828