Learn More
Medchemexpress LLC Camizestrant | 2222844-89-3 | 99.3% | 476.51 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY136255100MG
Encompass_Preferred
Camizestrant (AZD-9833) is a potent and orally active estrogen receptor (ER) antagonist used for the study of ER+ HER2-advanced breast cancer.
  • Potent and orally active estrogen receptor (ER) antagonist.
  • Exhibits anti-tumour efficacy in human parental MCF7 mice xenograft in a dose-dependent manner.
  • Decreases tumor growth in a dose-dependent manner in mice, showing almost complete tumor growth inhibition at doses >10 mg/kg.
  • Available in various quantities as a solid or in solution (10 mM in DMSO).
Catalog No. 50-003-51358